Learn More
Aviva Systems Biology Corp GALNT4 Antibody - middle region (ARP45527_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45527_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GALNT4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GALNT4. Alternate names: GALNAC-T4, GalNAcT4, GALNACT4ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-59047