Learn More
Cayman Chemical GalntamInhydrobromIde 50mg
SDP
Catalog No. 502913749
Encompass_Preferred
An alkaloid with AChE inhibitory and nAChR potentiating activities; selectively inhibits AChE over BChE (IC50s = 636 and 8,404 nM, respectively); potentiates ACh-induced currents in HEK293 cells expressing human α4β2 subunit-containing nAChRs at 0.5 µM; decreases escape latency and path length in the Morris water maze in the APP23 transgenic mouse model of Alzheimer's disease at 1.3 mg/kg per day
Catalog No. 50-291-3749 Supplier Cayman Chemical Supplier No. 3524750 MG
An alkaloid with AChE inhibitory and nAChR potentiating activities; selectively inhibits AChE over BChE (IC50s = 636 and 8,404 nM, respectively); potentiates ACh-induced currents in HEK293 cells expressing human α4β2 subunit-containing nAChRs at 0.5 µM; decreases escape latency and path length in the Morris water maze in the APP23 transgenic mouse model of Alzheimer's disease at 1.3 mg/kg per day