Learn More
Cayman Chemical GalocatechIn 10mg
SDP
Catalog No. 502966138
Encompass_Preferred
An epimer of (−)-EGC and potent antioxidant; significantly inhibits the proliferation of HCT116 and SW480 cancer cells; inhibits the aggregation of amyloid β peptide in vitro (IC50 = 17.5 µM)
Catalog No. 50-296-6138 Supplier Cayman Chemical Supplier No. 1694410 MG
An epimer of (−)-EGC and potent antioxidant; significantly inhibits the proliferation of HCT116 and SW480 cancer cells; inhibits the aggregation of amyloid β peptide in vitro (IC50 = 17.5 µM)