Learn More
Cayman Chemical (+)-Gallocatechin, Purity: ≥95%, CAS: 970-73-0, 50 mg
SDP
Catalog No. 502935287
Encompass_Preferred
A polyphenol and flavonoid that has diverse biological activities; inhibits adherence of P. gingivalis onto human buccal epithelial cells by >50% at 250 μg/ml; has antimutagenic properties in UV-irradiated E. coli cells; inhibits cell death induced by D-galactosamine and TNF-α in primary cultured mouse hepatocytes by 29.9% at 80 μM; inhibits HCT116 colorectal cancer cell proliferation by 57% at 100 μM
Catalog No. 50-293-5287 Supplier Cayman Chemical Supplier No. 2512550 MG
A polyphenol and flavonoid that has diverse biological activities; inhibits adherence of P. gingivalis onto human buccal epithelial cells by >50% at 250 μg/ml; has antimutagenic properties in UV-irradiated E. coli cells; inhibits cell death induced by D-galactosamine and TNF-α in primary cultured mouse hepatocytes by 29.9% at 80 μM; inhibits HCT116 colorectal cancer cell proliferation by 57% at 100 μM