Learn More
Cayman Chemical (−)-Gallocatechin, Purity: ≥98%, CAS: 3371-27-5, 5 mg
SDP
Catalog No. 502966990
Encompass_Preferred
An epimer of (−)-EGC and potent antioxidant; significantly inhibits the proliferation of HCT116 and SW480 cancer cells; inhibits the aggregation of amyloid β peptide in vitro (IC50 = 17.5 µM)
Catalog No. 50-296-6990 Supplier Cayman Chemical Supplier No. 169445 MG
An epimer of (−)-EGC and potent antioxidant; significantly inhibits the proliferation of HCT116 and SW480 cancer cells; inhibits the aggregation of amyloid β peptide in vitro (IC50 = 17.5 µM)