Learn More
Cayman Chemical Galoflavn 10mg
SDP
Catalog No. 502971693
Encompass_Preferred
An LDH inhibitor (Kis = 5.46 and 15.06 µM, for the A and B isoforms, respectively) that prevents proliferation of many different cancer cells by blocking glycolysis and ATP production
Catalog No. 50-297-1693 Supplier Cayman Chemical Supplier No. 1484610 MG
An LDH inhibitor (Kis = 5.46 and 15.06 µM, for the A and B isoforms, respectively) that prevents proliferation of many different cancer cells by blocking glycolysis and ATP production