Learn More
Aviva Systems Biology Corp GALR2 Antibody - C-terminal region (ARP61395_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61395_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GALR2. It was validated on Western Blot. . Alternate names: GALNR2, MGC125983, MGC125984ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-287-363