Learn More
Medchemexpress LLC HY-13226 10mg Medchemexpress, Galunisertib CAS:700874-72-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1322610MG
Encompass_Preferred
Medchemexpress, HY-13226 10mg Galunisertib CAS:700874-72-2 Galunisertib (LY2157299) is a oral and selective TGF-β receptor type I (TGF-βRI) kinase inhibitor with an IC50 of 56 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8962