Learn More
Medchemexpress LLC HY-13226 50mg , Galunisertib CAS:700874-72-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1322650MG
Encompass_Preferred
HY-13226 50mg Galunisertib CAS: 700874-72-2 Galunisertib (LY2157299) is an oral and selective TGF-β receptor type I (TGF-βRI) kinase inhibitor with an IC50 of 56 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9687