Learn More
MSC Miller SkyGrip Horizontal Lifeline System, Galvanized Steel, 60 ft., 2-Worker Capacity, 1 ea.
Catalog No. 5000508043
Encompass
This temporary horizontal lifeline system is designed for use in steel applications, providing a reliable safety solution for up to 2 workers. The 60 ft. (18.3 m) system is constructed from durable, corrosion-resistant, zinc-plated steel and is designed for efficient installation and enhanced productivity in fall protection settings.
  • Compact and lightweight design facilitates easier handling, installation, and storage.
  • Patent-pending above-the-beam adjustment enables single-person installation and dismantle.
  • Energy absorber maintains consistent system forces during a fall to minimize fall clearance.
  • Integrated tension indicator ensures proper system tensioning.
  • Stanchions serve as single-point anchorages.
  • Fits beam flange widths from 4 in. to 18 in. and thicknesses up to 2-1/4 in.
  • Meets ANSI A10.32, CSA, and OSHA safety standards.
Catalog No. 50-005-08043 Supplier MSC Supplier No. 30422448
This temporary horizontal lifeline system is designed for use in steel applications, providing a reliable safety solution for up to 2 workers. The 60 ft. (18.3 m) system is constructed from durable, corrosion-resistant, zinc-plated steel and is designed for efficient installation and enhanced productivity in fall protection settings.
  • Compact and lightweight design facilitates easier handling, installation, and storage.
  • Patent-pending above-the-beam adjustment enables single-person installation and dismantle.
  • Energy absorber maintains consistent system forces during a fall to minimize fall clearance.
  • Integrated tension indicator ensures proper system tensioning.
  • Stanchions serve as single-point anchorages.
  • Fits beam flange widths from 4 in. to 18 in. and thicknesses up to 2-1/4 in.
  • Meets ANSI A10.32, CSA, and OSHA safety standards.