Learn More
MSC Quantum Storage Wire Shelf, Galvanized Steel, 14 in. x 72 in., 800 lb. Capacity, 1 ea.
Catalog No. 5000515160
Encompass
This wire shelf provides reliable storage for metalworking and industrial applications. Designed for durability, this shelf features a galvanized steel construction that is resistant to rust build-up, ensuring long-term utility in demanding environments.
  • Constructed from 18 gauge galvanized steel
  • Features double layer construction
  • Equipped with an edge on the border of the shelf to contain spills
  • Load capacity of 800 lb.
  • Dimensions measure 14 in. x 72 in. x 2 in.
Catalog No. 50-005-15160 Supplier MSC Supplier No. 91530600
This wire shelf provides reliable storage for metalworking and industrial applications. Designed for durability, this shelf features a galvanized steel construction that is resistant to rust build-up, ensuring long-term utility in demanding environments.
  • Constructed from 18 gauge galvanized steel
  • Features double layer construction
  • Equipped with an edge on the border of the shelf to contain spills
  • Load capacity of 800 lb.
  • Dimensions measure 14 in. x 72 in. x 2 in.