Learn More
Vikan™ Galvanized Steel Undercarriage
Catalog No. 17227669 Shop All Remco Products
Change view
Click to view available options
Color:
Steel
Catalog No. Color
17-227-669 Steel
1 options
Catalog No. 17-227-669 Supplier Vikan™ Supplier No. 6913
Only null left
Add to Cart
Add to Cart
Galvanized Steel Undercarriage is commonly used for meat, seafood, poultry, commercial fishing, confectionery, bakery, and grain applications.
  • Galvanized steel
  • Low undercarriage
  • Features caster wheels that are impervious to meat fat and bearings equipped with grease fittings
  • The zinc coating on the steel keeps the undercarriage free from rust, even in very moist environments
  • A low undercarriage is a great solution when mobility is required
Specifications
Product Type Undercarriage
Color Steel
Material Galvanized Steel
Quantity 1
Dimensions (L x W x H) 42.5 x 21.6 x 9.4 in.