Learn More
Pickering Laboratories Inc Simulated Lung Fluid, Gamble's solution, Stabilized, each (200 mL/bottle)
SDP
Catalog No. NC1873835
Encompass_Preferred
Respiratory tract fluid is a complex, still not well-characterized mixture containing salts, locally secreted proteins, lipids, surfactants and other compounds. Respiratory fluid is also not homogeneous and has unique regional characteristics. Gamble's solution represents the interstitial fluid deep within the lung and is used to simulate different lung conditions. It is used in pulmonary drug delivery studies as well as in studies of particles inhalation effects. Citrate is used in Gamble's solution instead of proteins to avoid foaming and acetate instead of organic acids. Gamble's solution has a pH of 7.4. Inquire about other simulated lung fluid formulations
Catalog No. NC1873835 Supplier Pickering Laboratories Inc Supplier No. 17000808
Respiratory tract fluid is a complex, still not well-characterized mixture containing salts, locally secreted proteins, lipids, surfactants and other compounds. Respiratory fluid is also not homogeneous and has unique regional characteristics. Gamble's solution represents the interstitial fluid deep within the lung and is used to simulate different lung conditions. It is used in pulmonary drug delivery studies as well as in studies of particles inhalation effects. Citrate is used in Gamble's solution instead of proteins to avoid foaming and acetate instead of organic acids. Gamble's solution has a pH of 7.4. Inquire about other simulated lung fluid formulations