Learn More
Medchemexpress LLC HY-N5024 5mg Medchemexpress, Gambogenic acid CAS:173932-75-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN50245MG
Encompass_Preferred
Medchemexpress, HY-N5024 5mg Gambogenic acid CAS:173932-75-7 Gambogenic acid is an active ingredient in gamboge, with anticancer activity. Gambogenic acid acts as an effective inhibitor of EZH2, specifically and covalently binds to Cys668 within the EZH2-SET domain, and induces EZH2 ubiquitination. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0439