Learn More
Apexbio Technology LLC Gambogic Acid 2752-65-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063133
Encompass_Preferred
Gambogic Acid (CAS 2752-65-0) is a natural cytotoxic compound known to induce apoptosis through interactions with multiple apoptotic regulatory proteins It primarily targets anti-apoptotic Bcl-2 family members including Bcl-XL Bcl-2 Bcl-W Bcl-B Bfl-1 and Mcl-1 competitively binding to their BH3 domains and disrupting their suppression of mitochondrial pro-apoptotic factors It activates caspase-driven apoptotic pathways with EC50 values between 0 78-1 64 M Gambogic Acid demonstrates selective toxicity against cancer cell lines such as human gastric carcinoma MGC-803 cells and U266 multiple myeloma cells and inhibits tumor growth and angiogenesis in various in vivo xenograft models suggesting potential value for cancer biology research
Catalog No. 50-000-63133 Supplier Apexbio Technology LLC Supplier No. A34245.1
Add to Cart
Add to Cart
Gambogic Acid (CAS 2752-65-0) is a natural cytotoxic compound known to induce apoptosis through interactions with multiple apoptotic regulatory proteins It primarily targets anti-apoptotic Bcl-2 family members including Bcl-XL Bcl-2 Bcl-W Bcl-B Bfl-1 and Mcl-1 competitively binding to their BH3 domains and disrupting their suppression of mitochondrial pro-apoptotic factors It activates caspase-driven apoptotic pathways with EC50 values between 0 78-1 64 M Gambogic Acid demonstrates selective toxicity against cancer cell lines such as human gastric carcinoma MGC-803 cells and U266 multiple myeloma cells and inhibits tumor growth and angiogenesis in various in vivo xenograft models suggesting potential value for cancer biology research