Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gambogic acid | 2752-65-0 | 98.8% | 628.75 g/mol | C38H44O8 | 10 MG
SDP

Catalog No. 5000295851
Encompass_Preferred

Gambogic acid is a natural xanthonoid compound isolated from the resin of Garcinia hanburyi that functions as a Bcl-2 family inhibitor and is supplied for research use.

  • CAS number: 2752-65-0.
  • Molecular formula: C38H44O8.
  • Molecular weight: 628.75 g/mol.
  • Purity: 98.8% (typical).
  • Appearance: yellow to orange solid.
  • Storage: powder at -20°C (long term) or 4°C (short term); in solution at -80°C (long term).
  • Available in solid and solution formats, including 10 MG solid and solution concentrations for research applications.

Catalog No. 50-002-95851 Supplier Medchemexpress LLC Supplier No. HYN008710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Gambogic acid is a natural xanthonoid compound isolated from the resin of Garcinia hanburyi that functions as a Bcl-2 family inhibitor and is supplied for research use.

  • CAS number: 2752-65-0.
  • Molecular formula: C38H44O8.
  • Molecular weight: 628.75 g/mol.
  • Purity: 98.8% (typical).
  • Appearance: yellow to orange solid.
  • Storage: powder at -20°C (long term) or 4°C (short term); in solution at -80°C (long term).
  • Available in solid and solution formats, including 10 MG solid and solution concentrations for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.