Learn More
Bioworld Gamborg B5 Medium, 50 L
Catalog No. 50254605
Encompass_Preferred
Gamborg B5 MediumContains Micronutrients and Macronutrients as described by Gamborg, et al. (1968)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of mediumWorking Concentration: 3.1 g/LpH: 3.5–4.5Soluble in waterThis product is sold for laboratory research use only
Catalog No. 50-254-605 Supplier Bioworld Supplier No. 306300283
Gamborg B5 MediumContains Micronutrients and Macronutrients as described by Gamborg, et al. (1968)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of mediumWorking Concentration: 3.1 g/LpH: 3.5–4.5Soluble in waterThis product is sold for laboratory research use only