Learn More
Bioworld Gamborg B5 Medium w/ Vitamins, 50 L
Catalog No. 50254608
Encompass_Preferred
Gamborg B5 MediumContains Micronutrients, Macronutrients, and Vitamins as described by Gamborg, et al. (1968)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, 50L, or 100L of mediumWorking Concentration: 3.21 g/L This product is sold for laboratory research use only.
Catalog No. 50-254-608 Supplier Bioworld Supplier No. 306300294
Gamborg B5 MediumContains Micronutrients, Macronutrients, and Vitamins as described by Gamborg, et al. (1968)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, 50L, or 100L of mediumWorking Concentration: 3.21 g/L This product is sold for laboratory research use only.