Learn More
Bioworld Gamborg B5 Vitamin Concentration (Syringe Vials), 100 mL
Catalog No. 50254602
Encompass_Preferred
Gamborg B5 Vitamin Concentration Contains Vitamins as described by Gamborg, et al. (1968)Package includes 10 prepared, filtered, and sterilized ampules of 10 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: myo-Inositol: 100.00 mg/mLNicotinic Acid: 1.00 mg/mLPyridoxine HCl: 1.00 mg/mLThiamine HCl: 10.00 mg/mL
Catalog No. 50-254-602 Supplier Bioworld Supplier No. 306300261
Gamborg B5 Vitamin Concentration Contains Vitamins as described by Gamborg, et al. (1968)Package includes 10 prepared, filtered, and sterilized ampules of 10 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: myo-Inositol: 100.00 mg/mLNicotinic Acid: 1.00 mg/mLPyridoxine HCl: 1.00 mg/mLThiamine HCl: 10.00 mg/mL