Learn More
Bioworld Gamborg B5 Vitamin Mix, 100 mL
Catalog No. 50254603
Encompass_Preferred
Gamborg B5 Vitamin Mix Contains Vitamins as described by Gamborg, et al. (1968)Package includes 11.20 g vitamins to prepare 100 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: myo-Inositol: 100.00 mg/LNicotinic Acid: 1.00 mg/LPyridoxine HCl: 1.00 mg/LThiamine HCl: 10.00 mg/L
Catalog No. 50-254-603 Supplier Bioworld Supplier No. 306300271
Gamborg B5 Vitamin Mix Contains Vitamins as described by Gamborg, et al. (1968)Package includes 11.20 g vitamins to prepare 100 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: myo-Inositol: 100.00 mg/LNicotinic Acid: 1.00 mg/LPyridoxine HCl: 1.00 mg/LThiamine HCl: 10.00 mg/L