Learn More
PHYTOTECHNOLOGY LABS GAMBORG BASAL SALT MIXTUR 100L
Catalog No. NC1789700
Encompass_Preferred
This medium was developed for the initiation and growth of soybean cell suspensions. This medium contains no Ammonium Nitrate; it does contain Ammonium Sulfate and increased levels of Potassium Nitrate. Concentrations of NH4+ over 2 mM inhibited cell growth.
Catalog No. NC1789700 Supplier PHYTOTECHNOLOGY LABS Supplier No. G768100L
This medium was developed for the initiation and growth of soybean cell suspensions. This medium contains no Ammonium Nitrate; it does contain Ammonium Sulfate and increased levels of Potassium Nitrate. Concentrations of NH4+ over 2 mM inhibited cell growth.