Learn More
Research Products International Corp Gamborg's B5 Medium, Powder, Makes 1 Liter
SDP
Catalog No. 50213320
Encompass_Preferred
Basal salt mixture containing micro and macro elements as described by Gamborg, et al. (1968). Package contains 3.05 grams of powder per liter of medium.
Catalog No. 50-213-320 Supplier Research Products International Corp Supplier No. G201001.0
Add to Cart
Add to Cart
Basal salt mixture containing micro and macro elements as described by Gamborg, et al. (1968). Package contains 3.05 grams of powder per liter of medium.