Learn More
Research Products International Corp Gamborg's B5 Medium, Powder, Makes 50 Liters
SDP
Catalog No. 50213324
Encompass_Preferred
Basal salt mixture containing micro and macro elements as described by Gamborg, et al. (1968). Package contains 3.05 grams of powder per liter of medium.
Catalog No. 50-213-324 Supplier Research Products International Corp Supplier No. G2010050.0
Basal salt mixture containing micro and macro elements as described by Gamborg, et al. (1968). Package contains 3.05 grams of powder per liter of medium.