Learn More
Research Products International Corp Gamborg's B5 Medium with Vitamins, Powder, Makes 1 Liter
SDP
Catalog No. 50213325
Encompass_Preferred
Hygroscopic. Basal salt mixture containing micro and macro elements with vitamins as described by Gamborg, et. al. (1968). Package contains 3.16 grams of powder per liter of medium.
Catalog No. 50-213-325 Supplier Research Products International Corp Supplier No. G202001.0
Hygroscopic. Basal salt mixture containing micro and macro elements with vitamins as described by Gamborg, et. al. (1968). Package contains 3.16 grams of powder per liter of medium.