Learn More
Research Products International Corp Gamborg's B5 Medium Vitamin Mixture, 28 Grams of Powder, Makes 250 Milliliters of Solution
SDP
Catalog No. 50213345
Encompass_Preferred
Powdered mixture as described by Gamborg, et. al. (1968). Package contains powder vitamins to prepare of a 1000X vitamin stock solution. Use 1 milliliter vitamin stock solution to prepare 1 liter medium of final vitamin concentration.
Catalog No. 50-213-345 Supplier Research Products International Corp Supplier No. G370100.25
Powdered mixture as described by Gamborg, et. al. (1968). Package contains powder vitamins to prepare of a 1000X vitamin stock solution. Use 1 milliliter vitamin stock solution to prepare 1 liter medium of final vitamin concentration.