Learn More
PHYTOTECHNOLOGY LABS GAMBORGS B5 MEDIUM 100L
Catalog No. NC1789705
Encompass_Preferred
This medium was developed for the initiation and growth of soybean cell suspensions. This medium contains no Ammonium Nitrate; it does contain Ammonium Sulfate and increased levels of Potassium Nitrate. Concentrations of NH4+ over 2 mM inhibited cell growth.
Catalog No. NC1789705 Supplier PHYTOTECHNOLOGY LABS Supplier No. G398100L
This medium was developed for the initiation and growth of soybean cell suspensions. This medium contains no Ammonium Nitrate; it does contain Ammonium Sulfate and increased levels of Potassium Nitrate. Concentrations of NH4+ over 2 mM inhibited cell growth.