Learn More
Medchemexpress LLC Gamcemetinib | 1887069-10-4 | 99.1% | C22H20ClN5O3S | 10MG
SDP
Catalog No. 5000261481
Encompass_Preferred
Gamcemetinib is a covalent, irreversible inhibitor of MAP kinase-activated protein kinase-2 (MK2) used as a research compound for in vitro and preclinical studies. It is characterized by submicromolar activity in biochemical and cell-based assays and is supplied as a solid or as a DMSO solution for laboratory use.
  • Potent, covalent MK2 inhibitor with low-nanomolar to submicromolar activity.
  • High purity suitable for research use (≈99.1%).
  • Available as multiple solid quantities and as a 10 mM solution in DMSO for convenience.
  • Soluble in DMSO; enhanced solubility with warming and sonication.
  • Defined storage recommendations for powder and solution to preserve stability.
Catalog No. 50-002-61481 Supplier Medchemexpress LLC Supplier No. HY13950410MG
Gamcemetinib is a covalent, irreversible inhibitor of MAP kinase-activated protein kinase-2 (MK2) used as a research compound for in vitro and preclinical studies. It is characterized by submicromolar activity in biochemical and cell-based assays and is supplied as a solid or as a DMSO solution for laboratory use.
  • Potent, covalent MK2 inhibitor with low-nanomolar to submicromolar activity.
  • High purity suitable for research use (≈99.1%).
  • Available as multiple solid quantities and as a 10 mM solution in DMSO for convenience.
  • Soluble in DMSO; enhanced solubility with warming and sonication.
  • Defined storage recommendations for powder and solution to preserve stability.