Learn More
Cayman Chemical GamIthromycIn 50mg
SDP
Catalog No. 502986584
Encompass_Preferred
An antibiotic; active against macrolide-resistant R. equi and S. zooepidemicus (MIC90s = 1 and 0.125 μg/ml, respectively); reduces the number of pneumonic lesions in a bovine M. haemolytica challenge model at a dose of 6 mg/kg
Catalog No. 50-298-6584 Supplier Cayman Chemical Supplier No. 2338950 MG
An antibiotic; active against macrolide-resistant R. equi and S. zooepidemicus (MIC90s = 1 and 0.125 μg/ml, respectively); reduces the number of pneumonic lesions in a bovine M. haemolytica challenge model at a dose of 6 mg/kg