Learn More
Medchemexpress LLC HY-108365 5mg Medchemexpress, Gamithromycin CAS:145435-72-9 Purity:>98%
SDP
Catalog No. 501968541
Encompass_Preferred
Medchemexpress, HY-108365 5mg Gamithromycin CAS:145435-72-9 Gamithromycin is an antimicrobial agent which can inhibit the growth of MmmSC strains B237 and Tan8 with MICs of 0.00012 and 0.00006 μg/mL, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8541 Supplier Medchemexpress LLC Supplier No. HY1083655MG
Medchemexpress, HY-108365 5mg Gamithromycin CAS:145435-72-9 Gamithromycin is an antimicrobial agent which can inhibit the growth of MmmSC strains B237 and Tan8 with MICs of 0.00012 and 0.00006 μg/mL, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.