Learn More
Medchemexpress LLC Gamitrinib TPP hexafluorophosphate | 1131626-47-5 | 99.7% | 1036.03 | C52H65F6N3O8P2 | 10 MG
SDP
Catalog No. 5000247391
Encompass_Preferred
Gamitrinib TPP hexafluorophosphate is a mitochondria-targeted HSP90 inhibitor that selectively accumulates in the mitochondrial matrix and disrupts mitochondrial Hsp90 function. It is used in biochemical and cell-based cancer research to induce mitochondrial dysfunction and mitophagy and to probe mitochondrial protein homeostasis. The compound is supplied as a high-purity, pale purple solid suitable for in vitro studies.
  • Selectively targets mitochondria via a triphenylphosphonium moiety.
  • Inhibits mitochondrial HSP90 ATPase activity to disrupt protein folding.
  • Demonstrates anticancer activity in cell-based models.
  • High purity suitable for research applications.
  • Soluble in DMSO; insoluble in water.
  • Stable when stored under recommended low-temperature conditions.
Catalog No. 50-002-47391 Supplier Medchemexpress LLC Supplier No. HY102007A10MG
Gamitrinib TPP hexafluorophosphate is a mitochondria-targeted HSP90 inhibitor that selectively accumulates in the mitochondrial matrix and disrupts mitochondrial Hsp90 function. It is used in biochemical and cell-based cancer research to induce mitochondrial dysfunction and mitophagy and to probe mitochondrial protein homeostasis. The compound is supplied as a high-purity, pale purple solid suitable for in vitro studies.
  • Selectively targets mitochondria via a triphenylphosphonium moiety.
  • Inhibits mitochondrial HSP90 ATPase activity to disrupt protein folding.
  • Demonstrates anticancer activity in cell-based models.
  • High purity suitable for research applications.
  • Soluble in DMSO; insoluble in water.
  • Stable when stored under recommended low-temperature conditions.