Learn More
Medchemexpress LLC Gamitrinib TPP hexafluorophosphate | 1131626-47-5 | MFCD31715433 | 99.7% | 1,036.03 g/mol | C52H65F6N3O8P2 | 50 MG
SDP
Catalog No. 5000247415
Encompass_Preferred
Gamitrinib TPP hexafluorophosphate is a mitochondria-targeted HSP90 inhibitor used in preclinical research to selectively disrupt mitochondrial HSP90 function and impair tumor bioenergetics. The triphenylphosphonium (TPP) moiety directs the compound to the mitochondrial matrix, increasing mitochondrial accumulation and cytotoxic activity in cell and animal cancer models.
  • Mitochondria-targeted HSP90 inhibitor
  • Targets mitochondrial bioenergetics and induces cancer cell death
  • Enhanced mitochondrial accumulation via triphenylphosphonium moiety
  • Reported high chemical purity (≈99.7%)
  • Provided as a solid for preclinical research use
Catalog No. 50-002-47415 Supplier Medchemexpress LLC Supplier No. HY102007A50MG
Add to Cart
Add to Cart
Gamitrinib TPP hexafluorophosphate is a mitochondria-targeted HSP90 inhibitor used in preclinical research to selectively disrupt mitochondrial HSP90 function and impair tumor bioenergetics. The triphenylphosphonium (TPP) moiety directs the compound to the mitochondrial matrix, increasing mitochondrial accumulation and cytotoxic activity in cell and animal cancer models.
  • Mitochondria-targeted HSP90 inhibitor
  • Targets mitochondrial bioenergetics and induces cancer cell death
  • Enhanced mitochondrial accumulation via triphenylphosphonium moiety
  • Reported high chemical purity (≈99.7%)
  • Provided as a solid for preclinical research use