Learn More
Medchemexpress LLC Gamitrinib TPP hexafluorophosphate | 1131626-47-5 | MFCD31715433 | 99.7% | 1036.03 | C52H65F6N3O8P2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY102007A25MG
Encompass_Preferred
Gamitrinib TPP hexafluorophosphate is a mitochondria-targeted HSP90 inhibitor that selectively accumulates in the mitochondrial matrix and interferes with mitochondrial Hsp90 function, showing anticancer activity in preclinical studies. It couples an Hsp90 ATPase inhibitory module with a triphenylphosphonium mitochondrial-targeting moiety and is supplied for laboratory research use.
  • Mitochondria-targeted HSP90 inhibitor with anticancer activity.
  • Selective accumulation in the mitochondrial matrix for targeted action.
  • High purity suitable for research applications (≈99.7%).
  • Supplied as a solid; soluble in DMSO for biological assays.
  • Chemical formula C52H65F6N3O8P2; molecular weight 1036.03.
  • Intended for research use only; not for human or veterinary use.
  • Packaged in a 25 mg quantity for small-scale experiments.
Catalog No. 50-002-47414