Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gamma-[(6-Azidohexyl)-Imido]-Atp Sodium Salt

Catalog No. 8085201MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-852-01MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 80-852-01MG Supplier Sigma Aldrich Supplier No. 8085201MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lee, et al., reported a non-radioactive version of in vitro phosphorylation where +a-[(6-Azidohexyl)-imido]-ATP (compound 6) was successfully used instead of +a-32P-modified ATP to phosphorylate GST-tagged recombinant p27kip1 with protein kinase cdk2. The phosphorylated, azide-modified protein substrate can be labeled with alkynes of biotin or fluorescent dyes using Cu(I)-catalyzed click chemistry or DBCO-containing biotin or fluorescent dyes using Cu(I)-free click chemistry.The purchase of this product for use in applications relating to copper catalyzed azide-alkyne cycloaddition chemistry (Click Chemistry) includes a limited, nontransferable license to intellectual property owned by The Scripps Research Institute to use this product solely for internal non-commercial research activities and specifically excludes clinical, therapeutic, or diagnostic use in humans or animals and manufacturing purposes. Information regarding a license for commercial use in Click Chemistry may be obtained directly from The Scripps Research Institute, 10550 N. Torrey Pines Rd., La Jolla, CA 92037, or by contacting 858-784-8140 or click@scripps.edu.

Specifications

Melting Point 108°C to 110°C
Molecular Formula C21H27N5O5 -+ xH2O
Linear Formula C21H27N5O5 -+ xH2O
MDL Number MFCD00149318
Quantity 50 mg
Molecular Weight (g/mol) 429.47 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.