Learn More
TARGETMOL CHEMICALS INC GAMMA-ACETYLENIC GABA HY 25MG
SDP
Catalog No. 502576817
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (gamma)-Acetylenic GABA hydrochloride is a GABA transaminase inhibitor. purity: 99%
Catalog No. 50-257-6817 Supplier TARGETMOL CHEMICALS INC Supplier No. T40659L25MG
Also available in 5 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (gamma)-Acetylenic GABA hydrochloride is a GABA transaminase inhibitor. purity: 99%