Learn More
Medchemexpress LLC Gamma Adaptin Antibody (YA2993) | 50 UL
SDP
Catalog No. 5000443196
Encompass_Preferred
Adaptins are crucial components of clathrin-coated vesicles, facilitating the transport of ligand-receptor complexes. This protein belongs to the gamma-adaptin family, which forms heterotetrameric adaptor complexes vital for promoting the formation of clathrin-coated pits and vesicles.
  • Important for clathrin-coated vesicle function.
  • Transports ligand-receptor complexes from plasma membrane or trans-Golgi network to lysosomes.
  • Belongs to the adaptin family of proteins.
  • Adaptins, with medium and small subunits, form adaptor complexes.
  • Promotes formation of clathrin-coated pits and vesicles.
  • Encoded protein is a gamma-adaptin.
  • Two transcript variants encoding different isoforms found.
Catalog No. 50-004-43196 Supplier Medchemexpress LLC Supplier No. HYP8324850UL
Adaptins are crucial components of clathrin-coated vesicles, facilitating the transport of ligand-receptor complexes. This protein belongs to the gamma-adaptin family, which forms heterotetrameric adaptor complexes vital for promoting the formation of clathrin-coated pits and vesicles.
  • Important for clathrin-coated vesicle function.
  • Transports ligand-receptor complexes from plasma membrane or trans-Golgi network to lysosomes.
  • Belongs to the adaptin family of proteins.
  • Adaptins, with medium and small subunits, form adaptor complexes.
  • Promotes formation of clathrin-coated pits and vesicles.
  • Encoded protein is a gamma-adaptin.
  • Two transcript variants encoding different isoforms found.