Learn More
CPC Scientific H-Arg-Leu-Arg-Phe-Asp-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific BACP005A
Encompass
SEQUENCE: H-Arg-Leu-Arg-Phe-Asp-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: RLRFDMOLECULAR FORMULA: C31H51N11O8MOLECULAR WEIGHT: 705.8STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: Bag Cell Factor, gammaRESEARCH AREA: HormonalREFERENCES:R.H. Scheller et al., Cell, 32, 7 (1983)SKU(s): BACP-005A
Catalog No. 50-210-8838