Learn More
Selleck Chemical LLC (3-Carboxypropyl)trimethylammonium chloride S3361-25mg
SDP
Catalog No. 503108998
Encompass_Preferred
(3-Carboxypropyl)trimethylammonium chloride ( -Butyrobetaine BB hydrochloride) is an intermediary metabolite by gut microbes from dietary L-carnitine in mice
Catalog No. 50-310-8998 Supplier Selleck Chemical LLC Supplier No. S336125MG
(3-Carboxypropyl)trimethylammonium chloride ( -Butyrobetaine BB hydrochloride) is an intermediary metabolite by gut microbes from dietary L-carnitine in mice