Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gamma Catenin Antibody (YA3801) | 82 kDa | 10 UL
SDP

Catalog No. 5000449193
Encompass_Preferred

The gamma Catenin Antibody (YA3801) is a mouse-derived, non-conjugated IgG1 monoclonal antibody designed to target gamma Catenin. Gamma Catenin, also known as Catenin-y, is a common junctional plaque protein critical for cytoskeletal organization and cellular function. It plays a central role in desmosomes and intermediate junctions, influencing VE-cadherin function and potentially linking cadherins to the actin cytoskeleton.

  • Targets gamma Catenin
  • Host: Mouse
  • Monoclonal antibody
  • Non-conjugated
  • Immunogen: Purified recombinant fragment of human JUP aa 534-740
  • Molecular weight: 82 kDa
  • Human reactivity
  • Isotype: IgG1
  • Formulated in PBS with 0.05% sodium azide
  • Affinity purified
  • Suitable for WB, IHC-P, ICC/IF, and ELISA applications
  • Store at -20°C for 1 year, avoid freeze/thaw cycles

Catalog No. 50-004-49193 Supplier Medchemexpress LLC Supplier No. HYP8410410UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The gamma Catenin Antibody (YA3801) is a mouse-derived, non-conjugated IgG1 monoclonal antibody designed to target gamma Catenin. Gamma Catenin, also known as Catenin-y, is a common junctional plaque protein critical for cytoskeletal organization and cellular function. It plays a central role in desmosomes and intermediate junctions, influencing VE-cadherin function and potentially linking cadherins to the actin cytoskeleton.

  • Targets gamma Catenin
  • Host: Mouse
  • Monoclonal antibody
  • Non-conjugated
  • Immunogen: Purified recombinant fragment of human JUP aa 534-740
  • Molecular weight: 82 kDa
  • Human reactivity
  • Isotype: IgG1
  • Formulated in PBS with 0.05% sodium azide
  • Affinity purified
  • Suitable for WB, IHC-P, ICC/IF, and ELISA applications
  • Store at -20°C for 1 year, avoid freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.