Learn More
Medchemexpress LLC gamma Catenin Antibody (YA420) | 82 kDa | 50 UL
SDP
Catalog No. 5000396049
Encompass_Preferred
The gamma Catenin Antibody (YA420) is a rabbit-derived, non-conjugated IgG monoclonal antibody specifically targeting gamma Catenin. This protein is a crucial cytoplasmic component found in submembranous plaques of both desmosomes and intermediate junctions, forming distinct complexes with cadherins. As a member of the catenin family, it features a characteristic armadillo repeat motif. Mutations in the gene encoding gamma Catenin have been linked to Naxos disease.
  • Suitable for Western blot (WB) applications with a dilution ratio of 1:500-1:1000.
  • Applicable for immunohistochemistry-paraffin (IHC-P) at a dilution of 1:50-1:100.
  • Supports immunohistochemistry-frozen (IHC-F) with a dilution range of 1:50-1:100.
  • Compatible with immunocytochemistry/immunofluorescence (ICC/IF) at 1:50-1:200 dilution.
  • Can be used for multiplex immunohistochemistry (mIHC) with a 1:300 dilution.
Catalog No. 50-003-96049 Supplier Medchemexpress LLC Supplier No. HYP8068350UL
The gamma Catenin Antibody (YA420) is a rabbit-derived, non-conjugated IgG monoclonal antibody specifically targeting gamma Catenin. This protein is a crucial cytoplasmic component found in submembranous plaques of both desmosomes and intermediate junctions, forming distinct complexes with cadherins. As a member of the catenin family, it features a characteristic armadillo repeat motif. Mutations in the gene encoding gamma Catenin have been linked to Naxos disease.
  • Suitable for Western blot (WB) applications with a dilution ratio of 1:500-1:1000.
  • Applicable for immunohistochemistry-paraffin (IHC-P) at a dilution of 1:50-1:100.
  • Supports immunohistochemistry-frozen (IHC-F) with a dilution range of 1:50-1:100.
  • Compatible with immunocytochemistry/immunofluorescence (ICC/IF) at 1:50-1:200 dilution.
  • Can be used for multiplex immunohistochemistry (mIHC) with a 1:300 dilution.