Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gamma Catenin Antibody (YA420) | affinity purified | 82 kDa | 100UL
SDP

Catalog No. 5000395779
Encompass_Preferred

Gamma Catenin Antibody (YA420) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting gamma Catenin. This protein is a major cytoplasmic component common to submembranous plaques of both desmosomes and intermediate junctions. It forms distinct complexes with cadherins and desmosomal cadherins and is part of the catenin family, featuring an armadillo repeat motif. Mutations in this gene are associated with Naxos disease.

  • Host: Rabbit
  • Reactivity: Human
  • Conjugation: Non-conjugated
  • Research field: Signal transduction
  • Formulation: Supplied in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol and 0.05% BSA. Preservative: 0.01% Sodium azide
  • Appearance: Liquid
  • Shipping: Shipping with blue ice

Catalog No. 50-003-95779 Supplier Medchemexpress LLC Supplier No. HYP80683100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gamma Catenin Antibody (YA420) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting gamma Catenin. This protein is a major cytoplasmic component common to submembranous plaques of both desmosomes and intermediate junctions. It forms distinct complexes with cadherins and desmosomal cadherins and is part of the catenin family, featuring an armadillo repeat motif. Mutations in this gene are associated with Naxos disease.

  • Host: Rabbit
  • Reactivity: Human
  • Conjugation: Non-conjugated
  • Research field: Signal transduction
  • Formulation: Supplied in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol and 0.05% BSA. Preservative: 0.01% Sodium azide
  • Appearance: Liquid
  • Shipping: Shipping with blue ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.