Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gamma-Cyclodextrin

Catalog No. 779431100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-943-1100G 100 g
77-943-11KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 77-943-1100G Supplier Sigma Aldrich Supplier No. 779431100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Gamma-cyclodextrin (+a-CD) is a cyclic +a-1,4-glucan linked oligosaccharide that is widely used in the food and pharmaceutical industries. In drug delivery, +a-CD can be used as a complexing agent because it forms an inclusion complex with a drug molecule. The complex of cyclodextrin increases the aqueous solubility and dissolution rate of the drug, which is useful for the delivery of the medical agent to a biological system.+a-Cyclodextrin can be used as a precursor for the synthesis of: Octakis-(2,3-di-O-methyl-6-O-carboxymethyl)-+a-cyclodextrin sodium salt (ODMCM). ODMCM is used in capillary electrophoresis as a chiral resolving agent.Water-soluble cyclodextrin thioethers derivatives for the transportation of hydrophobic drugs.Cavasol is a registered trademark of Wacker Chemie AG

Specifications

CAS 56-89-3
Melting Point >240°C
Molecular Formula C6H12N2O4S2
Linear Formula [-SCH2CH(NH2)CO2H]2
MDL Number MFCD00064228
Quantity 100 g
Synonym Cavamax(R) W8; gamma-Cyclodextrin; Cyclomaltooctaose; Cyclooctaamylose; Schardinger gamma-Dextrin
Molecular Weight (g/mol) 240.3 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.