Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gamma-Cyclodextrin

Catalog No. 7796601KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-966-01KG 1 kg
77-966-0100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 77-966-01KG Supplier Sigma Aldrich Supplier No. 7796601KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

+a-Cyclodextrin (+a-CD), a water-soluble cyclic oligosaccharide consisting of eight +a-(1,4)-linked glucopyranose subunits, can be used in a wide range of applications including:Synthesis of renewable and biocompatible metal-organic frameworks.Formation of inclusion complexes with a variety of guest molecules to increase their solubility in water and other polar solvents.Synthesis of +a-CD based nanogels and dendrimers as carriers and stabilizers for drug delivery applications.Cavamax is a registered trademark of Wacker Chemie AG

Specifications

CAS 17465-86-0
Molecular Formula C48H80O40
Linear Formula C48H80O40
Quantity 1 kg
MDL Number MFCD00009595
Synonym Cavamax(R) W8; Cyclomaltooctaose; Cyclooctaamylose; Schardinger gamma-Dextrin
Molecular Weight (g/mol) 1297.12 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.