Learn More
Sigma Aldrich Fine Chemicals Biosciences gamma Cyclodextrin powder1G
Catalog No. 502902887
Encompass
Cyclodextrins belongs to the family of cyclic 1 4-glucans and is made of glucose units. This molecule has a shape of a hollow cone. It possesses a hydrophilic external and hydrophobic internal surface because of which cyclodextrins can form inclusion complexes.³-Cyclodextrin is extensively used in food and pharmaceutical industries. Cyclodextrins are obtained by the enzymatic conversion of starch/starch derivatives by cyclodextrin glycosyltransferase.
Catalog No. 50-290-2887 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C49301G
Cyclodextrins belongs to the family of cyclic 1 4-glucans and is made of glucose units. This molecule has a shape of a hollow cone. It possesses a hydrophilic external and hydrophobic internal surface because of which cyclodextrins can form inclusion complexes.³-Cyclodextrin is extensively used in food and pharmaceutical industries. Cyclodextrins are obtained by the enzymatic conversion of starch/starch derivatives by cyclodextrin glycosyltransferase.