Learn More
Medchemexpress LLC Gamma-D-glutamylglycine | 6729-55-1 | 99.2% | 204.18 g·mol⁻¹ | C7H12N2O5 | 10 MG
SDP
Catalog No. 5000211336
Encompass_Preferred
gamma-DGG (gamma-D-glutamylglycine) is a competitive AMPA receptor blocker used in neuroscience research to reduce AMPA-mediated synaptic currents. Supplied as a white to off-white solid and as a solution, it is used for in vitro and in vivo experiments requiring reversible AMPA antagonism and controlled stock preparation.
Catalog No. 50-002-11336 Supplier Medchemexpress LLC Supplier No. HY10078510MG
gamma-DGG (gamma-D-glutamylglycine) is a competitive AMPA receptor blocker used in neuroscience research to reduce AMPA-mediated synaptic currents. Supplied as a white to off-white solid and as a solution, it is used for in vitro and in vivo experiments requiring reversible AMPA antagonism and controlled stock preparation.