Learn More
Medchemexpress LLC Gamma-D-glutamylglycine | 6729-55-1 | 99.2% | 204.18 g·mol⁻¹ | C7H12N2O5 | 50 MG
SDP
Catalog No. 5000212396
Encompass_Preferred
gamma-DGG is a reversible, competitive AMPA receptor blocker used in neuropharmacology research to probe AMPA receptor-mediated synaptic transmission. It produces concentration-dependent blockade and reduces miniature excitatory postsynaptic current (mEPSC) amplitude, indicating a postsynaptic site of action.
  • Competitive AMPA receptor antagonist for studying synaptic transmission
  • Reversible action allows recovery after washout
  • Concentration-dependent blockade suitable for dose-response experiments
  • Reduces mEPSC amplitude, useful for postsynaptic mechanism studies
  • High purity and stable solid form for reliable experimental results
Catalog No. 50-002-12396 Supplier Medchemexpress LLC Supplier No. HY10078550MG
gamma-DGG is a reversible, competitive AMPA receptor blocker used in neuropharmacology research to probe AMPA receptor-mediated synaptic transmission. It produces concentration-dependent blockade and reduces miniature excitatory postsynaptic current (mEPSC) amplitude, indicating a postsynaptic site of action.
  • Competitive AMPA receptor antagonist for studying synaptic transmission
  • Reversible action allows recovery after washout
  • Concentration-dependent blockade suitable for dose-response experiments
  • Reduces mEPSC amplitude, useful for postsynaptic mechanism studies
  • High purity and stable solid form for reliable experimental results