Learn More
Medchemexpress LLC Gamma-D-glutamylglycine | 6729-55-1 | MFCD00063109 | 99.2% | 204.18 g·mol⁻1 | C7H12N2O5 | 100 MG
SDP
Catalog No. 5000212158
Encompass_Preferred
gamma-DGG (gamma-D-glutamylglycine) is a low-affinity, rapidly dissociating competitive antagonist of AMPA-type ionotropic glutamate receptors used in neuroscience research to probe AMPAR-mediated synaptic currents and receptor pharmacology.
  • Low-affinity competitive antagonist for AMPA receptors.
  • Rapid dissociation kinetics suitable for studying synaptic transmission.
  • Useful in electrophysiology to distinguish receptor contributions.
  • Reported high purity for reliable experimental use.
  • Available in small milligram pack sizes for laboratory studies.
Catalog No. 50-002-12158 Supplier Medchemexpress LLC Supplier No. HY100785100MG
gamma-DGG (gamma-D-glutamylglycine) is a low-affinity, rapidly dissociating competitive antagonist of AMPA-type ionotropic glutamate receptors used in neuroscience research to probe AMPAR-mediated synaptic currents and receptor pharmacology.
  • Low-affinity competitive antagonist for AMPA receptors.
  • Rapid dissociation kinetics suitable for studying synaptic transmission.
  • Useful in electrophysiology to distinguish receptor contributions.
  • Reported high purity for reliable experimental use.
  • Available in small milligram pack sizes for laboratory studies.