Learn More
Sigma Aldrich Fine Chemicals Biosciences gamma-Decalactone 98 fCC f1KG
Catalog No. 502899490
Encompass
I3-Decalactone the main volatile aroma compound found in peaches can be used as a flavoring agent in the food industry due to its distinct peach odor. It is also found in strawberries and apricots.
Catalog No. 50-289-9490 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W2360041KGK
I3-Decalactone the main volatile aroma compound found in peaches can be used as a flavoring agent in the food industry due to its distinct peach odor. It is also found in strawberries and apricots.