Learn More
Medchemexpress LLC HY-100785 5mg Medchemexpress, gamma-DGG CAS:6729-55-1 Purity:>98%
SDP
Catalog No. 501931948
Encompass_Preferred
Medchemexpress, HY-100785 5mg gamma-DGG CAS:6729-55-1 gamma-DGG is a competitive AMPA receptor blocker. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1948 Supplier Medchemexpress LLC Supplier No. HY1007855MG
Medchemexpress, HY-100785 5mg gamma-DGG CAS:6729-55-1 gamma-DGG is a competitive AMPA receptor blocker. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.