Learn More
Bachem H-Glu(Gln-OH)-OH
SDP
Supplier:  Bachem 4001245.0250
Encompass_Preferred
H-Glu(Gln-OH)-OH, 10148-81-9, C10H17N3O6, Mr 275.26, 250mg. H-g-Glu-Gln-OH. g-Glu-Gln and further kokumi-active g-glutamyl dipeptides have been isolated from mature Gouda cheese. These branched dipeptides are responsible for the complex taste of the cheese.
Catalog No. NC1747795