Learn More
Medchemexpress LLC Gamma-glutamylcysteine | 636-58-8 | 98.5% | 250.27 | 10 MG
SDP
Catalog No. 5000436717
Encompass_Preferred
Gamma-glutamylcysteine (γ-Glutamylcysteine) is a dipeptide containing cysteine and glutamic acid. It serves as a precursor to glutathione (GSH) and acts as a cofactor for glutathione peroxidase (GPx), helping to increase GSH levels in the body.
  • Precursor to glutathione (GSH)
  • Cofactor for glutathione peroxidase (GPx) to increase GSH levels
  • Increases activity of antioxidant enzymes like superoxide dismutase (SOD)
  • Leads to significant increases in total antioxidant capacity (TAC)
  • Reduces GSSG/GSH ratio
  • Upregulates anti-inflammatory cytokine IL-10
  • Reduces pro-inflammatory cytokines such as TNF-α and IL-6
  • Attenuates changes in metalloproteinase activity in oligomeric Aβ40-treated astrocytes
Catalog No. 50-004-36717 Supplier Medchemexpress LLC Supplier No. HY11340210MG
Gamma-glutamylcysteine (γ-Glutamylcysteine) is a dipeptide containing cysteine and glutamic acid. It serves as a precursor to glutathione (GSH) and acts as a cofactor for glutathione peroxidase (GPx), helping to increase GSH levels in the body.
  • Precursor to glutathione (GSH)
  • Cofactor for glutathione peroxidase (GPx) to increase GSH levels
  • Increases activity of antioxidant enzymes like superoxide dismutase (SOD)
  • Leads to significant increases in total antioxidant capacity (TAC)
  • Reduces GSSG/GSH ratio
  • Upregulates anti-inflammatory cytokine IL-10
  • Reduces pro-inflammatory cytokines such as TNF-α and IL-6
  • Attenuates changes in metalloproteinase activity in oligomeric Aβ40-treated astrocytes